Learn More
Biomatik Corporation CTPS1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08344
Encompass_Preferred
The CTPS1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human CTP synthase 1 protein (79-388AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200. Alternative Names: CTP synthase 1, CTP synthetase 1, UTP--ammonia ligase 1, CTPS1, CTPS. UniProt: P17812. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7927