Learn More
Biomatik Corporation CYBB anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08347
Encompass_Preferred
The CYBB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cytochrome b-245 heavy chain protein (283-570AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: CGD91-phox, Cytochrome b (558) subunit beta, Cytochrome b558 subunit beta, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, Superoxide-generating NADPH oxidase, heavy chain subunit, gp91-1, gp91-phox, p22 phagocyte B-cytochrome, NOX2, CYBB. UniProt: P04839. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7930