Learn More
Biomatik Corporation CYP3A7 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08350
Encompass_Preferred
The CYP3A7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cytochrome P450 3A7 protein (1-200AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cytochrome P450 3A7, CYPIIIA7, Cytochrome P450-HFLA, CYP3A7. UniProt: P24462. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7933