Learn More
Biomatik Corporation CYSLTR1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08351
Encompass_Preferred
The CYSLTR1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cysteinyl leukotriene receptor 1 protein (298-337AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cysteinyl leukotriene receptor 1, CysLTR1, Cysteinyl leukotriene D4 receptor, LTD4 receptor, G-protein coupled receptor HG55, HMTMF81, CYSLTR1, CYSLT1. UniProt: Q9Y271. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7934