Learn More
Biomatik Corporation DAO anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08352
Encompass_Preferred
The DAO Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human D-amino-acid oxidase protein (1-347AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: D-amino-acid oxidase, DAAO, DAMOX, DAO, DAMOX. UniProt: P14920. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7935