Learn More
Biomatik Corporation BRINP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507936
Encompass_Preferred
The BRINP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human BMP/retinoic acid-inducible neural-specific protein 1 protein (20-320AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: BMP/retinoic acid-inducible neural-specific protein 1, Deleted in bladder cancer protein 1, BRINP1, DBC1, DBCCR1, FAM5A, IB3089A. UniProt: O60477. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7936 Supplier Biomatik Corporation Supplier No. CAC08353
The BRINP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human BMP/retinoic acid-inducible neural-specific protein 1 protein (20-320AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: BMP/retinoic acid-inducible neural-specific protein 1, Deleted in bladder cancer protein 1, BRINP1, DBC1, DBCCR1, FAM5A, IB3089A. UniProt: O60477. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.