Learn More
Biomatik Corporation RPS19BP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08354
Encompass_Preferred
Rps19 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPS19. The Rps19 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 40S Ribosomal Protein S19 Protein (1-145Aa). Applications: ELISA, WB, IHC. Target Synonyms: RPS19. Accession Number: P39019. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7937