Learn More
Biomatik Corporation DDAH2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507938
Encompass_Preferred
The DDAH2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human N (G),N (G)-dimethylarginine dimethylaminohydrolase 2 protein (1-285AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: N (G), N (G)-dimethylarginine dimethylaminohydrolase 2, DDAH-2, Dimethylarginine dimethylaminohydrolase 2, DDAHII, Dimethylargininase-2, Protein G6a, S-phase protein, DDAH2, DDAH, G6A, NG30. UniProt: O95865. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7938 Supplier Biomatik Corporation Supplier No. CAC08355
The DDAH2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human N (G),N (G)-dimethylarginine dimethylaminohydrolase 2 protein (1-285AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: N (G), N (G)-dimethylarginine dimethylaminohydrolase 2, DDAH-2, Dimethylarginine dimethylaminohydrolase 2, DDAHII, Dimethylargininase-2, Protein G6a, S-phase protein, DDAH2, DDAH, G6A, NG30. UniProt: O95865. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.