Learn More
Biomatik Corporation DDHD2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507939
Encompass_Preferred
The DDHD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Phospholipase DDHD2 protein (361-650AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Phospholipase DDHD2 (EC:3.1.1.-), DDHD2, KIAA0725, SAMWD1, DDHD domain-containing protein 2, KIAA0725p, SAM, WWE and DDHD domain-containing protein 1. UniProt: O94830. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7939 Supplier Biomatik Corporation Supplier No. CAC08356
The DDHD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Phospholipase DDHD2 protein (361-650AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Phospholipase DDHD2 (EC:3.1.1.-), DDHD2, KIAA0725, SAMWD1, DDHD domain-containing protein 2, KIAA0725p, SAM, WWE and DDHD domain-containing protein 1. UniProt: O94830. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.