Learn More
Biomatik Corporation MP68 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507940
Encompass_Preferred
Mp68 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MP68. The Mp68 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 6.8 Kda Mitochondrial Proteolipid Protein (1-58Aa). Applications: ELISA, WB. Target Synonyms: MP68. Accession Number: P56378. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7940 Supplier Biomatik Corporation Supplier No. CAC08357
Mp68 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MP68. The Mp68 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 6.8 Kda Mitochondrial Proteolipid Protein (1-58Aa). Applications: ELISA, WB. Target Synonyms: MP68. Accession Number: P56378. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.