Learn More
Biomatik Corporation DHRS12 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08360
Encompass_Preferred
The DHRS12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Dehydrogenase/reductase SDR family member 12 protein (1-271AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dehydrogenase/reductase SDR family member 12 (EC:1.1.-.-), DHRS12, SDR40C11, Short-chain dehydrogenase/reductase family 40C member 11. UniProt: A0PJE2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7943