Learn More
Biomatik Corporation RPS9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507947
Encompass_Preferred
Rps9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPS9. The Rps9 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 40S Ribosomal Protein S9 Protein (2-184Aa). Applications: ELISA, WB. Target Synonyms: RPS9. Accession Number: P46781. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7947 Supplier Biomatik Corporation Supplier No. CAC08364
Rps9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPS9. The Rps9 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 40S Ribosomal Protein S9 Protein (2-184Aa). Applications: ELISA, WB. Target Synonyms: RPS9. Accession Number: P46781. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.