Learn More
Biomatik Corporation DMKN anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08365
Encompass_Preferred
The DMKN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Dermokine protein (1-137AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Dermokine, DMKN, UNQ729/PRO1411, Epidermis-specific secreted protein SK30/SK89. UniProt: Q6E0U4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7948