Learn More
Biomatik Corporation DNAJA1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08366
Encompass_Preferred
The DNAJA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DnaJ homolog subfamily A member 1 protein (8-106AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:1000-1:2000, IF:1:50-1:500. Alternative Names: DnaJ homolog subfamily A member 1, DnaJ protein homolog 2, HSDJ, Heat shock 40 kDa protein 4, Heat shock protein J2, HSJ-2, Human DnaJ protein 2, hDj-2, DNAJA1, DNAJ2, HDJ2, HSJ2, HSPF4. UniProt: P31689. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7949