Learn More
Biomatik Corporation DNAJB5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08368
Encompass_Preferred
The DNAJB5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DnaJ homolog subfamily B member 5 protein (1-344AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200. Alternative Names: DnaJ homolog subfamily B member 5, Heat shock protein Hsp40-2, Heat shock protein Hsp40-3, Heat shock protein cognate 40, Hsc40, DNAJB5, HSC40. UniProt: O75953. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7951