Learn More
Biomatik Corporation DOK5 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08374
Encompass_Preferred
The DOK5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Docking protein 5 protein (1-198AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Downstream of tyrosine kinase 5 Insulin receptor substrate 6 DOK5 C20orf180. UniProt: Q9P104. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7957