Learn More
Biomatik Corporation DPM1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507958
Encompass_Preferred
The DPM1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Dolichol-phosphate mannosyltransferase subunit 1 protein (2-260AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dolichol-phosphate mannosyltransferase subunit 1, Dolichol-phosphate mannose synthase subunit 1, DPM synthase subunit 1, Dolichyl-phosphate beta-D-mannosyltransferase subunit 1, Mannose-P-dolichol synthase subunit 1, MPD synthase subunit 1, DPM1. UniProt: O60762. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7958 Supplier Biomatik Corporation Supplier No. CAC08375
The DPM1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Dolichol-phosphate mannosyltransferase subunit 1 protein (2-260AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dolichol-phosphate mannosyltransferase subunit 1, Dolichol-phosphate mannose synthase subunit 1, DPM synthase subunit 1, Dolichyl-phosphate beta-D-mannosyltransferase subunit 1, Mannose-P-dolichol synthase subunit 1, MPD synthase subunit 1, DPM1. UniProt: O60762. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.