Learn More
Biomatik Corporation DSCR9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08376
Encompass_Preferred
The DSCR9 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Down syndrome critical region protein 9 protein (1-149AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Down syndrome critical region protein 9, DSCR9. UniProt: P59020. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7959