Learn More
Biomatik Corporation DSG3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08377
Encompass_Preferred
The DSG3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Desmoglein-3 protein (70-615AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: 130 kDa pemphigus vulgaris antigen, PVA, DSG3, CDHF6. UniProt: P32926. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7960