Learn More
Biomatik Corporation EBI3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507963
Encompass_Preferred
The EBI3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Interleukin-27 subunit beta protein (21-227AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: Interleukin-27 subunit beta, IL-27 subunit beta, IL-27B, Epstein-Barr virus-induced gene 3 protein, EBV-induced gene 3 protein, EBI3, IL27B. UniProt: Q14213. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7963 Supplier Biomatik Corporation Supplier No. CAC08380
The EBI3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Interleukin-27 subunit beta protein (21-227AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: Interleukin-27 subunit beta, IL-27 subunit beta, IL-27B, Epstein-Barr virus-induced gene 3 protein, EBV-induced gene 3 protein, EBI3, IL27B. UniProt: Q14213. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.