Learn More
Biomatik Corporation ECEL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507964
Encompass_Preferred
The ECEL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Endothelin-converting enzyme-like 1 protein (425-775AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Endothelin-converting enzyme-like 1, Xce protein, ECEL1, XCE, UNQ2431/PRO4991. UniProt: O95672. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7964 Supplier Biomatik Corporation Supplier No. CAC08381
The ECEL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Endothelin-converting enzyme-like 1 protein (425-775AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Endothelin-converting enzyme-like 1, Xce protein, ECEL1, XCE, UNQ2431/PRO4991. UniProt: O95672. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.