Learn More
Biomatik Corporation EEF1D anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08383
Encompass_Preferred
The EEF1D Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Elongation factor 1-delta protein (2-281AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Antigen NY-CO-4 EEF1D EF1D. UniProt: P29692. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7966