Learn More
Biomatik Corporation ELANE anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507974
Encompass_Preferred
The ELANE Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Neutrophil elastase protein (30-267AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Bone marrow serine protease, Elastase-2, Human leukocyte elastase, HLE, Medullasin, PMN elastase, ELANE, ELA2. UniProt: P08246. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7974 Supplier Biomatik Corporation Supplier No. CAC08391
The ELANE Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Neutrophil elastase protein (30-267AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Bone marrow serine protease, Elastase-2, Human leukocyte elastase, HLE, Medullasin, PMN elastase, ELANE, ELA2. UniProt: P08246. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.