Learn More
Biomatik Corporation CELA3A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08392
Encompass_Preferred
The CELA3A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Chymotrypsin-like elastase family member 3A protein (29-270AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Elastase IIIA, Elastase-3A, Protease E, ELA3, ELA3A, CELA3A. UniProt: P09093. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7975