Learn More
Biomatik Corporation ELAVL4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08393
Encompass_Preferred
The ELAVL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ELAV-like protein 4 protein (1-150AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ELAV-like protein 4, ELAVL4, HUD, PNEM, Hu-antigen D, HuD, Paraneoplastic encephalomyelitis antigen HuD. UniProt: P26378. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7976