Learn More
Biomatik Corporation EMP3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08395
Encompass_Preferred
The EMP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Epithelial membrane protein 3 protein (25-65AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: Epithelial membrane protein 3, EMP-3, Hematopoietic neural membrane protein 1, HNMP-1, Protein YMP, EMP3, YMP. UniProt: P54852. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7978