Learn More
Biomatik Corporation ENC1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08396
Encompass_Preferred
The ENC1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Ectoderm-neural cortex protein 1 protein (1-230AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Ectoderm-neural cortex protein 1, ENC-1, ENC1, KLHL37, NRPB, PIG10, Kelch-like protein 37, Nuclear matrix protein NRP/B, p53-induced gene 10 protein. UniProt: O14682. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7979