Learn More
Biomatik Corporation ENTPD3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08399
Encompass_Preferred
The ENTPD3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ectonucleoside triphosphate diphosphohydrolase 3 protein (60-338AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Ectonucleoside triphosphate diphosphohydrolase 3 (EC:3.6.1.5), NTPDase 3, ENTPD3, CD39L3, CD39 antigen-like 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPDase 3, Ecto-ATPase 3, Ecto-apyrase 3, HB6. UniProt: O75355. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7982