Learn More
Biomatik Corporation ENTPD7 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08400
Encompass_Preferred
The ENTPD7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Ectonucleoside triphosphate diphosphohydrolase 7 protein (51-350AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:2000-1:10000, IF:1:50-1:200. Alternative Names: Ectonucleoside triphosphate diphosphohydrolase 7, NTPDase 7, Lysosomal apyrase-like protein 1, ENTPD7, LALP1. UniProt: Q9NQZ7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7983