Learn More
Biomatik Corporation EPHX2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08405
Encompass_Preferred
The EPHX2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Bifunctional epoxide hydrolase 2 (1-555AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Bifunctional epoxide hydrolase 2, Epoxide hydratase, Soluble epoxide hydrolase, SEH, Lipid-phosphate phosphatase, EPHX2. UniProt: P34913. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7988