Learn More
Biomatik Corporation ERLIN1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507991
Encompass_Preferred
The ERLIN1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Erlin-1 protein (27-346AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Erlin-1, ERLIN1, C10orf69, KE04, KEO4, SPFH1, Endoplasmic reticulum lipid raft-associated protein 1, Protein KE04, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1, SPFH domain-containing protein 1. UniProt: O75477. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7991 Supplier Biomatik Corporation Supplier No. CAC08408
The ERLIN1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Erlin-1 protein (27-346AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Erlin-1, ERLIN1, C10orf69, KE04, KEO4, SPFH1, Endoplasmic reticulum lipid raft-associated protein 1, Protein KE04, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1, SPFH domain-containing protein 1. UniProt: O75477. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.