Learn More
Biomatik Corporation ETHE1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507993
Encompass_Preferred
The ETHE1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Persulfide dioxygenase ETHE1, mitochondrial protein (8-254AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Persulfide dioxygenase ETHE1, mitochondrial (EC:1.13.11.182 Publications, ETHE1, HSCO, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, Sulfur dioxygenase ETHE1. UniProt: O95571. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7993 Supplier Biomatik Corporation Supplier No. CAC08410
The ETHE1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Persulfide dioxygenase ETHE1, mitochondrial protein (8-254AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Persulfide dioxygenase ETHE1, mitochondrial (EC:1.13.11.182 Publications, ETHE1, HSCO, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, Sulfur dioxygenase ETHE1. UniProt: O95571. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.