Learn More
Biomatik Corporation ETNK1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08411
Encompass_Preferred
The ETNK1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ethanolamine kinase 1 protein (90-258AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Ethanolamine kinase 1 (EC:2.7.1.82), EKI 1, ETNK1, EKI1. UniProt: Q9HBU6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7994