Learn More
Biomatik Corporation EXO1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08414
Encompass_Preferred
The EXO1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Exonuclease 1 protein (301-600AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Exonuclease 1, hExo1, Exonuclease I, hExoI, EXO1, EXOI, HEX1. UniProt: Q9UQ84. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7997