Learn More
Biomatik Corporation F2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08415
Encompass_Preferred
The F2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Prothrombin protein (44-622AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:2000-1:5000, IF:1:50-1:200. Alternative Names: Prothrombin (EC:3.4.21.5), F2, Coagulation factor II, Activation peptide fragment 1, Activation peptide fragment 2, Thrombin light chain, Thrombin heavy chain. UniProt: P00734. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7998