Learn More
Biomatik Corporation F8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508002
Encompass_Preferred
The F8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Coagulation factor VIII protein (1-216AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Coagulation factor VIII, Antihemophilic factor, AHF, Procoagulant component, F8, F8C. UniProt: P00451. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8002 Supplier Biomatik Corporation Supplier No. CAC08419
The F8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Coagulation factor VIII protein (1-216AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Coagulation factor VIII, Antihemophilic factor, AHF, Procoagulant component, F8, F8C. UniProt: P00451. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.