Learn More
Biomatik Corporation FAM127A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08424
Encompass_Preferred
The FAM127A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Protein FAM127A protein (1-113AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200. Alternative Names: Protein FAM127A, Mammalian retrotransposon derived protein 8C, FAM127A, CXX1, MAR8, MAR8C, MART8. UniProt: A6ZKI3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8007