Learn More
Biomatik Corporation FASLG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08428
Encompass_Preferred
The FASLG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Tumor necrosis factor ligand superfamily member 6 protein (103-281AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:50-1:200, IF:1:50-1:500. Alternative Names: Apoptosis antigen ligand, APTL, CD95 ligand, CD95-L, Fas antigen ligand, Fas ligand, FasL, CD178, Receptor-binding FasL ectodomain, Soluble Fas ligand, sFasL, APL, FasL ICD, SPPL2A-processed FasL form, SPA, FASLG, APT1LG1, CD95L, FASL, TNFSF6. UniProt: P48023. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8011