Learn More
Biomatik Corporation FBXL18 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08429
Encompass_Preferred
The FBXL18 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human F-box/LRR-repeat protein 18 protein (1-365AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-box and leucine-rich repeat protein 18, FBXL18, FBL18. UniProt: Q96ME1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8012