Learn More
Biomatik Corporation FCER1G anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08430
Encompass_Preferred
The FCER1G Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human High affinity immunoglobulin epsilon receptor subunit gammaprotein (45-86AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Fc receptor gamma-chain Fc-epsilon RI-gamma IgE Fc receptor subunit gamma FCER1G. UniProt: P30273. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8013