Learn More
Biomatik Corporation FDFT1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08433
Encompass_Preferred
The FDFT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Squalene synthase protein (2-417AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Squalene synthase, SQS, SS, FPP:FPP farnesyltransferase, Farnesyl-diphosphate farnesyltransferase, FDFT1. UniProt: P37268. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8016