Learn More
Biomatik Corporation FDXR anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08434
Encompass_Preferred
The FDXR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human NADPH:adrenodoxin oxidoreductase, mitochondrial protein (33-451AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: NADPH:adrenodoxin oxidoreductase, mitochondrial (EC:1.18.1.6), AR, Adrenodoxin reductase, FDXR, ADXR, Ferredoxin--NADP (+) reductase, Ferredoxin reductase. UniProt: P22570. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8017