Learn More
Biomatik Corporation FFAR2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08435
Encompass_Preferred
The FFAR2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Free fatty acid receptor 2 protein (277-330AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Free fatty acid receptor 2, G-protein coupled receptor 43, FFAR2, FFA2, GPCR43, GPR43. UniProt: O15552. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8018