Learn More
Biomatik Corporation FGF13 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08436
Encompass_Preferred
The FGF13 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Fibroblast growth factor 13 protein (1-245AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Fibroblast growth factor 13, FGF-13, FGF13, FHF2, Fibroblast growth factor homologous factor 2, FHF-2. UniProt: Q92913. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8019