Learn More
Biomatik Corporation FGFR1OP2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08437
Encompass_Preferred
The FGFR1OP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human FGFR1 oncogene partner 2 protein (1-172AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: FGFR1 oncogene partner 2, FGFR1OP2, HSPC123. UniProt: Q9NVK5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8020