Learn More
Biomatik Corporation FKBP3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08440
Encompass_Preferred
The FKBP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Peptidyl-prolyl cis-trans isomerase FKBP3 protein (2-224AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 25 kDa FK506-binding protein, FK506-binding protein 3, Immunophilin FKBP25, Rapamycin-selective 25 kDa immunophilin, Rotamase, FKBP25, FKBP3. UniProt: Q00688. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8023