Learn More
Biomatik Corporation FLG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08443
Encompass_Preferred
The FLG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Filaggrin protein (3838-4061AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: FLG. UniProt: P20930. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8026