Learn More
Biomatik Corporation FMO4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08445
Encompass_Preferred
The FMO4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Dimethylaniline monooxygenase [N-oxide-forming] 4 protein (1-300AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dimethylaniline monooxygenase [N-oxide-forming] 4, Dimethylaniline oxidase 4, Hepatic flavin-containing monooxygenase 4, FMO 4, FMO4, FMO2. UniProt: P31512. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8028