Learn More
Biomatik Corporation FNTB anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08446
Encompass_Preferred
The FNTB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Protein farnesyltransferase subunit beta protein (1-300AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200, IP:1:200-1:2000. Alternative Names: Protein farnesyltransferase subunit beta, FTase-beta, CAAX farnesyltransferase subunit beta, Ras proteins prenyltransferase subunit beta, FNTB. UniProt: P49356. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8029