Learn More
Biomatik Corporation FPR2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08449
Encompass_Preferred
The FPR2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human N-formyl peptide receptor 2 protein (307-351AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: N-formyl peptide receptor 2, FPR2, FPRH1, FPRL1, LXA4R, FMLP-related receptor I, FMLP-R-I, Formyl peptide receptor-like 1, HM63, Lipoxin A4 receptor, LXA4 receptor, RFP. UniProt: P25090. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8032